Learn More
Biomatik Corporation Mouse Chymase 1, Mast Cell (CMA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03186
Encompass_Preferred
Mouse Chymase 1, Mast Cell (CMA1) ELISA Kit for detecting CMA1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21844. Gene ID: 17228. Target Synonyms: CYH; CYM; Alpha-chymase; Mast cell protease I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2110