Learn More
Biomatik Corporation Human Clara Cell Protein 16 (CC16) ELISA Kit, 96 tests
Catalog No. 501502112
Encompass_Preferred
The Human Clara Cell Protein 16 (CC16) ELISA Kit allows for quantitative determination of Clara Cell Protein 16 in Serum, Plasma, Tissue homogenates, Cell lysates, Lung lavage fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11684. Cross Activity: No significant cross-reactivity or interference between Clara Cell Protein 16 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2112 Supplier Biomatik Corporation Supplier No. EKU03199
The Human Clara Cell Protein 16 (CC16) ELISA Kit allows for quantitative determination of Clara Cell Protein 16 in Serum, Plasma, Tissue homogenates, Cell lysates, Lung lavage fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P11684. Cross Activity: No significant cross-reactivity or interference between Clara Cell Protein 16 and analogues was observed. Restriction: For Research Use Only.