Learn More
Biomatik Corporation Mouse Clara Cell Protein 16 (CC16) ELISA Kit, 96 tests
Catalog No. 501502113
Encompass_Preferred
Mouse Clara Cell Protein 16 (CC16) ELISA Kit for detecting CC16 in serum, plasma, lung lavage fluid, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06318. Gene ID: 22287. Target Synonyms: SCGB1A1; CC10; CCSP; UGB; UG; Uteroglobin; Uteroglobin; Urinary Protein 1; Clara Cell Secretory Protein; Secretoglobin Family 1A Member 1; Clara-Cell Specific 10-kD Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2113 Supplier Biomatik Corporation Supplier No. EKU03200
Mouse Clara Cell Protein 16 (CC16) ELISA Kit for detecting CC16 in serum, plasma, lung lavage fluid, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06318. Gene ID: 22287. Target Synonyms: SCGB1A1; CC10; CCSP; UGB; UG; Uteroglobin; Uteroglobin; Urinary Protein 1; Clara Cell Secretory Protein; Secretoglobin Family 1A Member 1; Clara-Cell Specific 10-kD Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.