Learn More
Biomatik Corporation Human Claudin 3 (CLDN3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03207
Encompass_Preferred
The Human Claudin 3 (CLDN3) ELISA Kit allows for quantitative determination of Claudin 3 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O15551. Cross Activity: No significant cross-reactivity or interference between Claudin 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2115