Learn More
Biomatik Corporation Rat Claudin 5 (CLDN5) ELISA Kit, 96 tests
Catalog No. 501502118
Encompass_Preferred
Rat Claudin 5 (CLDN5) ELISA Kit for detecting CLDN5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JKD6. Gene ID: 65131. Target Synonyms: AWAL; BEC1; CPETRL1; TMVCF; Transmembrane Protein Deleted In Velocardiofacial Syndrome; Transmembrane protein deleted in VCFS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2118 Supplier Biomatik Corporation Supplier No. EKU03214
Rat Claudin 5 (CLDN5) ELISA Kit for detecting CLDN5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JKD6. Gene ID: 65131. Target Synonyms: AWAL; BEC1; CPETRL1; TMVCF; Transmembrane Protein Deleted In Velocardiofacial Syndrome; Transmembrane protein deleted in VCFS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.