Learn More
Biomatik Corporation Human Circadian Locomoter Output Cycles Protein Kaput (CLOCK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03219
Encompass_Preferred
The Human Circadian Locomoter Output Cycles Protein Kaput (CLOCK) ELISA Kit allows for quantitative determination of Circadian Locomoter Output Cycles Protein Kaput in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O15516. Cross Activity: No significant cross-reactivity or interference between Circadian Locomoter Output Cycles Protein Kaput and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2119