Learn More
Biomatik Corporation Pig Cluster of Differentiation 14 (CD14) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03225
Encompass_Preferred
Pig Cluster Of Differentiation 14 (CD14) ELISA Kit for detecting CD14 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 6.25-400ng/ml. Sensitivity: 2.49ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A2SW51. Target Synonyms: Monocyte differentiation antigen CD14; Myeloid cell-specific leucine-rich glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2122