Learn More
Biomatik Corporation Rat Cluster of Differentiation 14 (CD14) ELISA Kit, 96 tests
Catalog No. 501502123
Encompass_Preferred
Rat Cluster Of Differentiation 14 (CD14) ELISA Kit for detecting CD14 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63691. Gene ID: 60350. Target Synonyms: Monocyte differentiation antigen CD14; Myeloid cell-specific leucine-rich glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2123 Supplier Biomatik Corporation Supplier No. EKU03226
Rat Cluster Of Differentiation 14 (CD14) ELISA Kit for detecting CD14 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63691. Gene ID: 60350. Target Synonyms: Monocyte differentiation antigen CD14; Myeloid cell-specific leucine-rich glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.