Learn More
Biomatik Corporation Human Cluster of Differentiation 19 (CD19) ELISA Kit, 96 tests
Catalog No. 501502124
Encompass_Preferred
The Human Cluster of Differentiation 19 (CD19) ELISA Kit allows for quantitative determination of Cluster of Differentiation 19 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.118 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P15391. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 19 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2124 Supplier Biomatik Corporation Supplier No. EKU03228
The Human Cluster of Differentiation 19 (CD19) ELISA Kit allows for quantitative determination of Cluster of Differentiation 19 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.118 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P15391. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 19 and analogues was observed. Restriction: For Research Use Only.