Learn More
Biomatik Corporation Human Cluster of Differentiation 200 (CD200) ELISA Kit, 96 tests
Catalog No. 501502125
Encompass_Preferred
The Human Cluster of Differentiation 200 (CD200) ELISA Kit allows for quantitative determination of Cluster of Differentiation 200 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 12.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41217. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 200 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2125 Supplier Biomatik Corporation Supplier No. EKU03231
The Human Cluster of Differentiation 200 (CD200) ELISA Kit allows for quantitative determination of Cluster of Differentiation 200 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2,000 pg/mL. Sensitivity: 12.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41217. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 200 and analogues was observed. Restriction: For Research Use Only.