Learn More
Biomatik Corporation Mouse Cluster of Differentiation 34 (CD34) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03237
Encompass_Preferred
Mouse Cluster Of Differentiation 34 (CD34) ELISA Kit for detecting CD34 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64314. Gene ID: 12490. Target Synonyms: Hematopoietic progenitor cell antigen CD34. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2128