Learn More
Biomatik Corporation Human Cluster of Differentiation 4 (CD4) ELISA Kit, 96 tests
Catalog No. 501502129
Encompass_Preferred
The Human Cluster of Differentiation 4 (CD4) ELISA Kit allows for quantitative determination of Cluster of Differentiation 4 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01730. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2129 Supplier Biomatik Corporation Supplier No. EKU03238
The Human Cluster of Differentiation 4 (CD4) ELISA Kit allows for quantitative determination of Cluster of Differentiation 4 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.064 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01730. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 4 and analogues was observed. Restriction: For Research Use Only.