Learn More
Biomatik Corporation Human Cluster of Differentiation 40 Ligand (CD40L) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03240
Encompass_Preferred
The Human Cluster of Differentiation 40 Ligand (CD40L) ELISA Kit allows for quantitative determination of Cluster of Differentiation 40 Ligand in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29965. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 40 Ligand and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2131