Learn More
Biomatik Corporation Rat Cluster of Differentiation 40 Ligand (CD40L) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03243
Encompass_Preferred
Rat Cluster Of Differentiation 40 Ligand (CD40L) ELISA Kit for detecting CD40L in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z2V2. Gene ID: 84349. Target Synonyms: CD154; TRAP; HIGM1; IGM; IMD3; TBAM; T-BAM; TNFSF5; Gp39; TNF Superfamily Member 5; Hyper-IgM Syndrome; TNF-Related Activation Protein; T-Cell B-Cell Activating Molecule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2132