Learn More
Biomatik Corporation Human Cluster of Differentiation 52 (CD52) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03244
Encompass_Preferred
The Human Cluster of Differentiation 52 (CD52) ELISA Kit allows for quantitative determination of Cluster of Differentiation 52 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P31358. Cross Activity: No significant cross-reactivity or interference between Cluster of Differentiation 52 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2133