Learn More
Biomatik Corporation Mouse Cluster of Differentiation 83 (CD83) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03247
Encompass_Preferred
Mouse Cluster Of Differentiation 83 (CD83) ELISA Kit for detecting CD83 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 17.88pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88324. Gene ID: 12522. Target Synonyms: BL11; HB15; Activated B Lymphocytes,Immunoglobulin; B-cell activation protein; Cell surface protein HB15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2136