Learn More
Biomatik Corporation Human Clusterin (CLU) ELISA Kit, 96 tests
Catalog No. 501502137
Encompass_Preferred
The Human Clusterin (CLU) ELISA Kit allows for quantitative determination of Clusterin in Serum, Plasma, Urine, Saliva, Tissue homogenates, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5-800 ng/mL. Sensitivity: 5.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10909. Cross Activity: No significant cross-reactivity or interference between Clusterin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2137 Supplier Biomatik Corporation Supplier No. EKU03249
The Human Clusterin (CLU) ELISA Kit allows for quantitative determination of Clusterin in Serum, Plasma, Urine, Saliva, Tissue homogenates, Cerebrospinal fluid, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5-800 ng/mL. Sensitivity: 5.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10909. Cross Activity: No significant cross-reactivity or interference between Clusterin and analogues was observed. Restriction: For Research Use Only.