Learn More
Biomatik Corporation Rat Clusterin (CLU) ELISA Kit, 96 tests
Catalog No. 501502139
Encompass_Preferred
Rat Clusterin (CLU) ELISA Kit for detecting CLU in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05371. Gene ID: 24854. Target Synonyms: CLU; TRPM2; KUB1; CLI; APOJ; SGP2; AAG4; APO-J; SP-40; Apolipoprotein J; Complement Lysis Inhibitor; 40,Sulfated Glycoprotein 2; Testosterone-Repressed Prostate Message 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2139 Supplier Biomatik Corporation Supplier No. EKU03251
Rat Clusterin (CLU) ELISA Kit for detecting CLU in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.37ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05371. Gene ID: 24854. Target Synonyms: CLU; TRPM2; KUB1; CLI; APOJ; SGP2; AAG4; APO-J; SP-40; Apolipoprotein J; Complement Lysis Inhibitor; 40,Sulfated Glycoprotein 2; Testosterone-Repressed Prostate Message 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.