Learn More
Biomatik Corporation Human Coagulation Factor II (F2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03256
Encompass_Preferred
The Human Coagulation Factor II (F2) ELISA Kit allows for quantitative determination of Coagulation Factor II in Serum. Species Reactivity: Homo sapiens (Human). Detection Range: 0.41-300 ng/mL. Sensitivity: 0.19 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00734. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor II and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2140