Learn More
Biomatik Corporation Human Coagulation Factor IX (F9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03259
Encompass_Preferred
The Human Coagulation Factor IX (F9) ELISA Kit allows for quantitative determination of Coagulation Factor IX in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5,000 pg/mL. Sensitivity: 32 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00740. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor IX and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2141