Learn More
Biomatik Corporation Human Coagulation Factor V (F5) ELISA Kit, 96 tests
Catalog No. 501502143
Encompass_Preferred
The Human Coagulation Factor V (F5) ELISA Kit allows for quantitative determination of Coagulation Factor V in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 61 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12259. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor V and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2143 Supplier Biomatik Corporation Supplier No. EKU03262
The Human Coagulation Factor V (F5) ELISA Kit allows for quantitative determination of Coagulation Factor V in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 61 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12259. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor V and analogues was observed. Restriction: For Research Use Only.