Learn More
Biomatik Corporation Mouse Coagulation Factor V (F5) ELISA Kit, 96 tests
Catalog No. 501502144
Encompass_Preferred
Mouse Coagulation Factor V (F5) ELISA Kit for detecting F5 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88783. Gene ID: 14067. Target Synonyms: FV; FVL; PCCF; Proaccelerin; Labile Factor; Factor V Leiden; Accelerator Globulin; Activated protein C cofactor; Proaccelerin, labile factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2144 Supplier Biomatik Corporation Supplier No. EKU03263
Mouse Coagulation Factor V (F5) ELISA Kit for detecting F5 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 50pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88783. Gene ID: 14067. Target Synonyms: FV; FVL; PCCF; Proaccelerin; Labile Factor; Factor V Leiden; Accelerator Globulin; Activated protein C cofactor; Proaccelerin, labile factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.