Learn More
Biomatik Corporation Rat Coagulation Factor V (F5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03264
Encompass_Preferred
Rat Coagulation Factor V (F5) ELISA Kit for detecting F5 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7TPK2. Gene ID: 304929. Target Synonyms: FV; FVL; PCCF; Proaccelerin; Labile Factor; Factor V Leiden; Accelerator Globulin; Activated protein C cofactor; Proaccelerin, labile factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2145