Learn More
Biomatik Corporation Mouse Coagulation Factor VII (F7) ELISA Kit, 96 tests
Catalog No. 501502147
Encompass_Preferred
Mouse Coagulation Factor VII (F7) ELISA Kit for detecting F7 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70375. Gene ID: 14068. Target Synonyms: FVII; SPCA; proconvertin; Cothromboplastin; Serum Prothrombin Conversation Accelerator; Stable Factor; Eptacog alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2147 Supplier Biomatik Corporation Supplier No. EKU03267
Mouse Coagulation Factor VII (F7) ELISA Kit for detecting F7 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70375. Gene ID: 14068. Target Synonyms: FVII; SPCA; proconvertin; Cothromboplastin; Serum Prothrombin Conversation Accelerator; Stable Factor; Eptacog alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.