Learn More
Biomatik Corporation Mouse Coagulation Factor VIII (F8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03271
Encompass_Preferred
Mouse Coagulation Factor VIII (F8) ELISA Kit for detecting F8 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.273ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06194. Gene ID: 14069. Target Synonyms: FVIII; AHF; HEMA; F8B; F8C; Antihemophilic Globulin; Hemophilia A; Anti-Hemophilic Factor A; FVIII Procoagulant Component; F8 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2150