Learn More
Biomatik Corporation Human Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03281
Encompass_Preferred
The Human Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA Kit allows for quantitative determination of Coagulation Factor XIII A1 Polypeptide in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.188-12 ng/mL. Sensitivity: 0.069 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00488. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor XIII A1 Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2155