Learn More
Biomatik Corporation Human Coenzyme Q10 Homolog B (COQ10B) ELISA Kit, 96 tests
Catalog No. 501502156
Encompass_Preferred
The Human Coenzyme Q10 Homolog B (COQ10B) ELISA Kit allows for quantitative determination of Coenzyme Q10 Homolog B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9H8M1. Cross Activity: No significant cross-reactivity or interference between Coenzyme Q10 Homolog B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2156 Supplier Biomatik Corporation Supplier No. EKU03287
The Human Coenzyme Q10 Homolog B (COQ10B) ELISA Kit allows for quantitative determination of Coenzyme Q10 Homolog B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9H8M1. Cross Activity: No significant cross-reactivity or interference between Coenzyme Q10 Homolog B and analogues was observed. Restriction: For Research Use Only.