Learn More
Biomatik Corporation Human Coiled Coil Domain Containing Protein 80 (CCDC80) ELISA Kit, 96 tests
Catalog No. 501502158
Encompass_Preferred
The Human Coiled Coil Domain Containing Protein 80 (CCDC80) ELISA Kit allows for quantitative determination of Coiled Coil Domain Containing Protein 80 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q76M96. Cross Activity: No significant cross-reactivity or interference between Coiled Coil Domain Containing Protein 80 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2158 Supplier Biomatik Corporation Supplier No. EKU03293
The Human Coiled Coil Domain Containing Protein 80 (CCDC80) ELISA Kit allows for quantitative determination of Coiled Coil Domain Containing Protein 80 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q76M96. Cross Activity: No significant cross-reactivity or interference between Coiled Coil Domain Containing Protein 80 and analogues was observed. Restriction: For Research Use Only.