Learn More
Biomatik Corporation Human Cold Inducible RNA Binding Protein (CIRBP) ELISA Kit, 96 tests
Catalog No. 501502159
Encompass_Preferred
The Human Cold Inducible RNA Binding Protein (CIRBP) ELISA Kit allows for quantitative determination of Cold Inducible RNA Binding Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14011. Cross Activity: No significant cross-reactivity or interference between Cold Inducible RNA Binding Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2159 Supplier Biomatik Corporation Supplier No. EKU03294
The Human Cold Inducible RNA Binding Protein (CIRBP) ELISA Kit allows for quantitative determination of Cold Inducible RNA Binding Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 35 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14011. Cross Activity: No significant cross-reactivity or interference between Cold Inducible RNA Binding Protein and analogues was observed. Restriction: For Research Use Only.