Learn More
Biomatik Corporation Human Collagen Triple Helix Repeat Containing Protein 1 (CTHRC1) ELISA Kit, 96 tests
Catalog No. 501502160
Encompass_Preferred
The Human Collagen Triple Helix Repeat Containing Protein 1 (CTHRC1) ELISA Kit allows for quantitative determination of Collagen Triple Helix Repeat Containing Protein 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.59 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96CG8. Cross Activity: No significant cross-reactivity or interference between Collagen Triple Helix Repeat Containing Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2160 Supplier Biomatik Corporation Supplier No. EKU03296
The Human Collagen Triple Helix Repeat Containing Protein 1 (CTHRC1) ELISA Kit allows for quantitative determination of Collagen Triple Helix Repeat Containing Protein 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.59 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96CG8. Cross Activity: No significant cross-reactivity or interference between Collagen Triple Helix Repeat Containing Protein 1 and analogues was observed. Restriction: For Research Use Only.