Learn More
Biomatik Corporation Mouse Collagen Type I (COL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03298
Encompass_Preferred
Mouse Collagen Type I (COL1) ELISA Kit for detecting COL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087; Q01149. Gene ID: 12842; 12843. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2162