Learn More
Biomatik Corporation Human Collagen Type I Alpha 1 (COL1a1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03300
Encompass_Preferred
The Human Collagen Type I Alpha 1 (COL1a1) ELISA Kit allows for quantitative determination of Collagen Type I Alpha 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13526. Cross Activity: No significant cross-reactivity or interference between Collagen Type I Alpha 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2164