Learn More
Biomatik Corporation Mouse Collagen Type I Alpha 1 (COL1a1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03301
Encompass_Preferred
Mouse Collagen Type I Alpha 1 (COL1a1) ELISA Kit for detecting COL1a1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087. Gene ID: 12842. Target Synonyms: COL-1A1; COL1-A1; COL1A-1; OI4; Collagen Alpha-1 (I) chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2165