Learn More
Biomatik Corporation Human Collagen Type IV (COL4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03321
Encompass_Preferred
The Human Collagen Type IV (COL4) ELISA Kit allows for quantitative determination of Collagen Type IV in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 ng/mL. Sensitivity: 3.1 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Collagen Type IV and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2170