Learn More
Biomatik Corporation Mouse Collagen Type IV Alpha 1 (COL4a1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03325
Encompass_Preferred
Mouse Collagen Type IV Alpha 1 (COL4a1) ELISA Kit for detecting COL4a1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02463. Gene ID: 12826. Target Synonyms: COL4-A1; Arresten; Collagen Alpha-1 (IV) chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2172