Learn More
Biomatik Corporation Human Collagen Type IV Alpha 3 (COL4a3) ELISA Kit, 96 tests
Catalog No. 501502173
Encompass_Preferred
The Human Collagen Type IV Alpha 3 (COL4a3) ELISA Kit allows for quantitative determination of Collagen Type IV Alpha 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q01955. Cross Activity: No significant cross-reactivity or interference between Collagen Type IV Alpha 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2173 Supplier Biomatik Corporation Supplier No. EKU03327
The Human Collagen Type IV Alpha 3 (COL4a3) ELISA Kit allows for quantitative determination of Collagen Type IV Alpha 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q01955. Cross Activity: No significant cross-reactivity or interference between Collagen Type IV Alpha 3 and analogues was observed. Restriction: For Research Use Only.