Learn More
Biomatik Corporation Human Collagen Type VII (COL7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03338
Encompass_Preferred
The Human Collagen Type VII (COL7) ELISA Kit allows for quantitative determination of Collagen Type VII in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.36 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q02388. Cross Activity: No significant cross-reactivity or interference between Collagen Type VII and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2177