Learn More
Biomatik Corporation Human Collagen Type XI Alpha 1 (COL11a1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03344
Encompass_Preferred
The Human Collagen Type XI Alpha 1 (COL11a1) ELISA Kit allows for quantitative determination of Collagen Type XI Alpha 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 39-2500 pg/mL. Sensitivity: 15 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12107. Cross Activity: No significant cross-reactivity or interference between Collagen Type XI Alpha 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2178