Learn More
Biomatik Corporation Human Collagen Type XV (COL15) ELISA Kit, 96 tests
Catalog No. 501502181
Encompass_Preferred
The Human Collagen Type XV (COL15) ELISA Kit allows for quantitative determination of Collagen Type XV in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P39059. Cross Activity: No significant cross-reactivity or interference between Collagen Type XV and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2181 Supplier Biomatik Corporation Supplier No. EKU03351
The Human Collagen Type XV (COL15) ELISA Kit allows for quantitative determination of Collagen Type XV in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P39059. Cross Activity: No significant cross-reactivity or interference between Collagen Type XV and analogues was observed. Restriction: For Research Use Only.