Learn More
Biomatik Corporation Human Collagen Type XVII (COL17) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03353
Encompass_Preferred
The Human Collagen Type XVII (COL17) ELISA Kit allows for quantitative determination of Collagen Type XVII in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.13 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UMD9. Cross Activity: No significant cross-reactivity or interference between Collagen Type XVII and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2182