Learn More
Biomatik Corporation Human Colony Stimulating Factor 2 Receptor Beta (CSF2Rb) ELISA Kit, 96 tests
Catalog No. 501502183
Encompass_Preferred
The Human Colony Stimulating Factor 2 Receptor Beta (CSF2Rb) ELISA Kit allows for quantitative determination of Colony Stimulating Factor 2 Receptor Beta in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P32927. Cross Activity: No significant cross-reactivity or interference between Colony Stimulating Factor 2 Receptor Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2183 Supplier Biomatik Corporation Supplier No. EKU03357
The Human Colony Stimulating Factor 2 Receptor Beta (CSF2Rb) ELISA Kit allows for quantitative determination of Colony Stimulating Factor 2 Receptor Beta in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P32927. Cross Activity: No significant cross-reactivity or interference between Colony Stimulating Factor 2 Receptor Beta and analogues was observed. Restriction: For Research Use Only.