Learn More
Biomatik Corporation Human Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) ELISA Kit, 96 tests
Catalog No. 501502184
Encompass_Preferred
The Human Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) ELISA Kit allows for quantitative determination of Colony Stimulating Factor 2, Granulocyte Macrophage in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04141. Cross Activity: No significant cross-reactivity or interference between Colony Stimulating Factor 2, Granulocyte Macrophage and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2184 Supplier Biomatik Corporation Supplier No. EKU03359
The Human Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) ELISA Kit allows for quantitative determination of Colony Stimulating Factor 2, Granulocyte Macrophage in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04141. Cross Activity: No significant cross-reactivity or interference between Colony Stimulating Factor 2, Granulocyte Macrophage and analogues was observed. Restriction: For Research Use Only.