Learn More
Biomatik Corporation Mouse Colony Stimulating Factor 2, Granulocyte Macrophage (GMCSF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03360
Encompass_Preferred
Mouse Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF) ELISA Kit for detecting GM-CSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1000pg/ml. Sensitivity: 0.56pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01587. Gene ID: 12981. Target Synonyms: CSF2; GMCSF; Sargramostim; Molgramostin; Granulocyte-Macrophage Colony Stimulating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2185