Learn More
Biomatik Corporation Mouse Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03363
Encompass_Preferred
Mouse Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit for detecting GCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.69-500pg/ml. Sensitivity: 0.29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09920. Gene ID: 12985. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2187