Learn More
Biomatik Corporation Rat Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit, 96 tests
Catalog No. 501502188
Encompass_Preferred
Rat Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit for detecting GCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97712. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2188 Supplier Biomatik Corporation Supplier No. EKU03364
Rat Colony Stimulating Factor 3, Granulocyte (GCSF) ELISA Kit for detecting GCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97712. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.