Learn More
Biomatik Corporation Human Complement C3 Convertase (C3c) ELISA Kit, 96 tests
Catalog No. 501502191
Encompass_Preferred
The Human Complement C3 Convertase (C3c) ELISA Kit allows for quantitative determination of Complement C3 Convertase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01024. Cross Activity: No significant cross-reactivity or interference between Complement C3 Convertase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2191 Supplier Biomatik Corporation Supplier No. EKU03371
The Human Complement C3 Convertase (C3c) ELISA Kit allows for quantitative determination of Complement C3 Convertase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01024. Cross Activity: No significant cross-reactivity or interference between Complement C3 Convertase and analogues was observed. Restriction: For Research Use Only.