Learn More
Biomatik Corporation Human Complement Component 3a (C3a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03387
Encompass_Preferred
The Human Complement Component 3a (C3a) ELISA Kit allows for quantitative determination of Complement Component 3a in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5,000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01024. Cross Activity: No significant cross-reactivity or interference between Complement Component 3a and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2197