Learn More
Biomatik Corporation Mouse Complement Component 3a (C3a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03388
Encompass_Preferred
Mouse Complement Component 3a (C3a) ELISA Kit for detecting C3a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01027. Gene ID: 12266. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2198