Learn More
Biomatik Corporation Pig Complement Component 3a (C3a) ELISA Kit, 96 tests
Catalog No. 501502199
Encompass_Preferred
Pig Complement Component 3a (C3a) ELISA Kit for detecting C3a in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500pg/ml. Sensitivity: 3.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01025. Gene ID: 397072. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2199 Supplier Biomatik Corporation Supplier No. EKU03389
Pig Complement Component 3a (C3a) ELISA Kit for detecting C3a in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500pg/ml. Sensitivity: 3.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01025. Gene ID: 397072. Restrictions: For Research Use Only. Not for use in diagnostic procedures.