Learn More
Biomatik Corporation Mouse Complement Component 4a (C4a) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03396
Encompass_Preferred
Mouse Complement Component 4a (C4a) ELISA Kit for detecting C4a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B9EIU2. Target Synonyms: C4a anaphylatoxin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2203